Learn More
Meso Scale Discovery R-PLEX Human -GST ASY
SDP
Catalog No. 5000082475
Encompass
The R-PLEX Human Alpha GST Assay Kit from Meso Scale Discovery measures alpha glutathione S-transferase (GSTA) in human serum plasma and cell culture supernatants This ECL immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive rapid GSTA detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours with a 3-minute read time per plate The assay range is 3 2-10 000 pg/mL with no cross-reactivity with other proteins
Catalog No. 50-000-82475 Supplier Meso Scale Discovery Supplier No. K151N1R2
The R-PLEX Human Alpha GST Assay Kit from Meso Scale Discovery measures alpha glutathione S-transferase (GSTA) in human serum plasma and cell culture supernatants This ECL immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive rapid GSTA detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are available in 3 5 hours with a 3-minute read time per plate The assay range is 3 2-10 000 pg/mL with no cross-reactivity with other proteins