Learn More
Meso Scale Discovery R-PLEX Human -GST ASYQP
SDP
Catalog No. 5000081182
Encompass
The R-PLEX Human Alpha GST Assay Kit from Meso Scale Discovery measures alpha glutathione S-transferase (GSTA1) in human serum plasma and other samples It uses MSD s electrochemiluminescent detection for high sensitivity a dynamic range of up to 5 logs and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals It quantifies alpha GST levels between 0 04 - 500 ng/mL with just 25 L of sample enabling accurate quantitation of the liver toxicity biomarker alpha GST
Catalog No. 50-000-81182 Supplier Meso Scale Discovery Supplier No. K151N1R22
The R-PLEX Human Alpha GST Assay Kit from Meso Scale Discovery measures alpha glutathione S-transferase (GSTA1) in human serum plasma and other samples It uses MSD s electrochemiluminescent detection for high sensitivity a dynamic range of up to 5 logs and low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals It quantifies alpha GST levels between 0 04 - 500 ng/mL with just 25 L of sample enabling accurate quantitation of the liver toxicity biomarker alpha GST