Learn More
Meso Scale Discovery R-PLEX Human Haptoglobin ASY
SDP
Catalog No. 5000078784
Encompass
The R-PLEX Human Haptoglobin Assay by Meso Scale Diagnostics is a quantitative immunoassay for detecting haptoglobin in human serum plasma and other fluids It uses MSD s electrochemiluminescent technology offering high sensitivity a broad dynamic range and low sample volume The assay is specific to haptoglobin with no cross-reactivity The kit includes a 96-well plate detection antibody calibrators and diluent Total assay time is 3 5 hours It s suitable for biomarker research and monitoring haptoglobin levels in disease states
Catalog No. 50-000-78784 Supplier Meso Scale Discovery Supplier No. K151YFR2
The R-PLEX Human Haptoglobin Assay by Meso Scale Diagnostics is a quantitative immunoassay for detecting haptoglobin in human serum plasma and other fluids It uses MSD s electrochemiluminescent technology offering high sensitivity a broad dynamic range and low sample volume The assay is specific to haptoglobin with no cross-reactivity The kit includes a 96-well plate detection antibody calibrators and diluent Total assay time is 3 5 hours It s suitable for biomarker research and monitoring haptoglobin levels in disease states