Learn More
Meso Scale Discovery R-PLEX Human Haptoglobin ASYQP
SDP
Catalog No. 5000079725
Encompass
The R-PLEX Human Haptoglobin Assay from Meso Scale Diagnostics is a quantitative immunoassay for the detection of haptoglobin in human serum plasma and other biological fluids This assay utilizes electrochemiluminescence technology on the MSD platform for high sensitivity and broad dynamic range The assay has a lower limit of detection of 0 08 ng/mL and an upper limit of 10 000 ng/mL The total assay time is 3 5 hours This assay provides a precise accurate high-throughput method to measure haptoglobin levels
Catalog No. 50-000-79725 Supplier Meso Scale Discovery Supplier No. K151YFR22
The R-PLEX Human Haptoglobin Assay from Meso Scale Diagnostics is a quantitative immunoassay for the detection of haptoglobin in human serum plasma and other biological fluids This assay utilizes electrochemiluminescence technology on the MSD platform for high sensitivity and broad dynamic range The assay has a lower limit of detection of 0 08 ng/mL and an upper limit of 10 000 ng/mL The total assay time is 3 5 hours This assay provides a precise accurate high-throughput method to measure haptoglobin levels