Learn More
Meso Scale Discovery R-PLEX Human HB-EGF ASY
SDP
Catalog No. 5000081148
Encompass
The R-PLEX Human HB-EGF Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Heparin-binding EGF-like growth factor (HB-EGF) in serum plasma cell culture supernatants and other biological fluids The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81148 Supplier Meso Scale Discovery Supplier No. K151H5R2
The R-PLEX Human HB-EGF Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Heparin-binding EGF-like growth factor (HB-EGF) in serum plasma cell culture supernatants and other biological fluids The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL