Learn More
Meso Scale Discovery R-PLEX Human HCC4CCL16 ASY
SDP
Catalog No. 5000078924
Encompass
The R-PLEX Human HCC-4 (CCL16) Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures the human chemokine CCL16 in serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents needed to perform the assay Results are obtained in as little as 3 5 hours This assay allows for the accurate quantitation of CCL16 levels in human samples
Catalog No. 50-000-78924 Supplier Meso Scale Discovery Supplier No. K151AATR2
The R-PLEX Human HCC-4 (CCL16) Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures the human chemokine CCL16 in serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents needed to perform the assay Results are obtained in as little as 3 5 hours This assay allows for the accurate quantitation of CCL16 levels in human samples