Learn More
Meso Scale Discovery R-PLEX Human hCG alpha ASY
SDP
Catalog No. 5000079577
Encompass
The R-PLEX Human HCG Alpha Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for measuring human chorionic gonadotropin alpha subunit (hCG ) in various biological matrices It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent Sensitivity is 0 05 mIU/mL with an upper limit of 1000 mIU/mL suitable for monitoring hCG levels and pregnancy testing
Catalog No. 50-000-79577 Supplier Meso Scale Discovery Supplier No. K151AGGR2
The R-PLEX Human HCG Alpha Assay Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for measuring human chorionic gonadotropin alpha subunit (hCG ) in various biological matrices It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent Sensitivity is 0 05 mIU/mL with an upper limit of 1000 mIU/mL suitable for monitoring hCG levels and pregnancy testing