Learn More
Meso Scale Discovery R-PLEX Human Hemojuvelin ASY
SDP
Catalog No. 5000078942
Encompass
The R-PLEX Human Hemojuvelin Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures hemojuvelin levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can be run on Meso Scale Diagnostic s SECTOR Imager 2400 or SECTOR S 600 plate readers and results are obtained in as little as 3 5 hours This product enables the quantification of hemojuvelin an iron regulatory protein for research applications
Catalog No. 50-000-78942 Supplier Meso Scale Discovery Supplier No. K151ACGR2
The R-PLEX Human Hemojuvelin Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures hemojuvelin levels in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can be run on Meso Scale Diagnostic s SECTOR Imager 2400 or SECTOR S 600 plate readers and results are obtained in as little as 3 5 hours This product enables the quantification of hemojuvelin an iron regulatory protein for research applications