Learn More
Meso Scale Discovery R-PLEX Human Hemojuvelin ASYQP
SDP
Catalog No. 5000079493
Encompass
The R-PLEX Human Hemojuvelin Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the simultaneous quantitative measurement of hemojuvelin in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a plate coated with capture antibodies a detection antibody solution calibrator solutions sample diluent wash buffer read buffer and adhesive plate seals The assay has a typical sensitivity of 0 3 ng/mL and an assay range from 0 5 - 1000 ng/mL
Catalog No. 50-000-79493 Supplier Meso Scale Discovery Supplier No. K151ACGR22
The R-PLEX Human Hemojuvelin Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the simultaneous quantitative measurement of hemojuvelin in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a plate coated with capture antibodies a detection antibody solution calibrator solutions sample diluent wash buffer read buffer and adhesive plate seals The assay has a typical sensitivity of 0 3 ng/mL and an assay range from 0 5 - 1000 ng/mL