Learn More
Meso Scale Discovery R-PLEX Human Hemopexin ASY
SDP
Catalog No. 5000080155
Encompass
The R-PLEX Human Hemopexin Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that quantitatively measures human hemopexin in serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrator The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 ng/mL
Catalog No. 50-000-80155 Supplier Meso Scale Discovery Supplier No. K1517FR2
The R-PLEX Human Hemopexin Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that quantitatively measures human hemopexin in serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrator The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 2500 ng/mL