Learn More
Meso Scale Discovery R-PLEX Human Hemopexin ASYQP
SDP
Catalog No. 5000080255
Encompass
The R-PLEX Human Hemopexin Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures hemopexin levels in human serum plasma and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies calibrators and reagents required to perform the assay Hemopexin is measured with high sensitivity and the kit provides a simple rapid way to quantify hemopexin levels from small sample volumes The assay has applications in biomarker research and disease monitoring
Catalog No. 50-000-80255 Supplier Meso Scale Discovery Supplier No. K1517FR22
The R-PLEX Human Hemopexin Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures hemopexin levels in human serum plasma and other biological samples The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies calibrators and reagents required to perform the assay Hemopexin is measured with high sensitivity and the kit provides a simple rapid way to quantify hemopexin levels from small sample volumes The assay has applications in biomarker research and disease monitoring