Learn More
Meso Scale Discovery R-PLEX Human Hepcidin ASY
SDP
Catalog No. 5000082272
Encompass
The Meso Scale Discovery R-PLEX Human Hepcidin Assay Kit is designed to quantitatively measure hepcidin levels in human serum plasma and other biological fluids This multiplex immunoassay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay requires an MSD instrument such as SECTOR Imager QuickPlex SQ 120 or QuickPlex SQ 120 Each kit provides reagents for 96 tests in a 96-well plate formatted assay
Catalog No. 50-000-82272 Supplier Meso Scale Discovery Supplier No. K151J1R2
The Meso Scale Discovery R-PLEX Human Hepcidin Assay Kit is designed to quantitatively measure hepcidin levels in human serum plasma and other biological fluids This multiplex immunoassay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay requires an MSD instrument such as SECTOR Imager QuickPlex SQ 120 or QuickPlex SQ 120 Each kit provides reagents for 96 tests in a 96-well plate formatted assay