Learn More
Meso Scale Discovery R-PLEX Human Hepcidin ASYQP
SDP
Catalog No. 5000082332
Encompass
The Meso Scale Discovery R-PLEX Human Hepcidin Assay Kit measures human hepcidin in serum plasma urine and other samples using electrochemiluminescence (ECL) technology It offers high sensitivity a broad dynamic range and quick results The kit includes a 96-well plate with capture and detection antibodies calibrators controls diluents buffers and seals It detects hepcidin from 0 064 ng/mL to 1000 ng/mL with just 50 L of sample in 3 5 hours providing a simple rapid method for quantifying hepcidin levels
Catalog No. 50-000-82332 Supplier Meso Scale Discovery Supplier No. K151J1R22
The Meso Scale Discovery R-PLEX Human Hepcidin Assay Kit measures human hepcidin in serum plasma urine and other samples using electrochemiluminescence (ECL) technology It offers high sensitivity a broad dynamic range and quick results The kit includes a 96-well plate with capture and detection antibodies calibrators controls diluents buffers and seals It detects hepcidin from 0 064 ng/mL to 1000 ng/mL with just 50 L of sample in 3 5 hours providing a simple rapid method for quantifying hepcidin levels