Learn More
Meso Scale Discovery R-PLEX Human HGF ASY
SDP
Catalog No. 5000080209
Encompass
The R-PLEX Human HGF Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human hepatocyte growth factor (HGF) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 10 000 pg/mL It requires 25 L of sample and can perform up to 80 tests in duplicate
Catalog No. 50-000-80209 Supplier Meso Scale Discovery Supplier No. K1514NR2
Add to Cart
Add to Cart
The R-PLEX Human HGF Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human hepatocyte growth factor (HGF) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator The assay has a typical sensitivity of 0 08 ng/mL and an upper limit of quantification of 10 000 pg/mL It requires 25 L of sample and can perform up to 80 tests in duplicate