Learn More
Meso Scale Discovery R-PLEX Human HGF ASYQP
SDP
Catalog No. 5000078847
Encompass
The R-PLEX Human HGF Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human hepatocyte growth factor (HGF) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-78847 Supplier Meso Scale Discovery Supplier No. K1514NR22
The R-PLEX Human HGF Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human hepatocyte growth factor (HGF) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL