Learn More
Meso Scale Discovery R-PLEX Human HSP27/HspB1 ASY
SDP
Catalog No. 5000078918
Encompass
The R-PLEX Human HSP27 (HSPB1) Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures human HSP27 protein levels in serum plasma cell culture supernatants cell and tissue lysates and other biological samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls This assay allows for the quantitative measurement of HSP27 levels with high precision and reproducibility
Catalog No. 50-000-78918 Supplier Meso Scale Discovery Supplier No. K151AGHR2
The R-PLEX Human HSP27 (HSPB1) Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures human HSP27 protein levels in serum plasma cell culture supernatants cell and tissue lysates and other biological samples This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and controls This assay allows for the quantitative measurement of HSP27 levels with high precision and reproducibility