Learn More
Meso Scale Discovery R-PLEX Human HSP27/HspB1 ASYQP
SDP
Catalog No. 5000080329
Encompass
The R-PLEX Human HSP27 (HSPB1) Assay is an ELISA kit for the quantitative measurement of HSP27 in human serum plasma cell culture supernatants cell and tissue lysates and other biological fluids It uses a quantitative sandwich immunoassay technique and comes with a 96-well strip plate that is pre-coated with antibodies specific for HSP27 The detection range for this assay is 15 6-1000 pg/mL with a sensitivity of 5 pg/mL This assay kit provides materials to run up to 40 samples in duplicate and takes around 3 hours to complete Overall the R-PLEX Human HSP27 Assay provides a sensitive high-throughput method for quantifying HSP27 levels in various human sample types
Catalog No. 50-000-80329 Supplier Meso Scale Discovery Supplier No. K151AGHR22
The R-PLEX Human HSP27 (HSPB1) Assay is an ELISA kit for the quantitative measurement of HSP27 in human serum plasma cell culture supernatants cell and tissue lysates and other biological fluids It uses a quantitative sandwich immunoassay technique and comes with a 96-well strip plate that is pre-coated with antibodies specific for HSP27 The detection range for this assay is 15 6-1000 pg/mL with a sensitivity of 5 pg/mL This assay kit provides materials to run up to 40 samples in duplicate and takes around 3 hours to complete Overall the R-PLEX Human HSP27 Assay provides a sensitive high-throughput method for quantifying HSP27 levels in various human sample types