Learn More
Meso Scale Discovery R-PLEX Human IgE ASYQP
SDP
Catalog No. 5000078409
Encompass
The R-PLEX Human IgE Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures IgE antibodies in human serum plasma or cell culture supernatant It uses electrochemiluminescence detection technology and provides quantitative results for 10 IgE biomarkers including IgE Total Ara h 1 Ara h 2 Ara h 3 Ara h 8 Ara h 9 Bos d 5 Bos d 8 Gal d 1 and Gal d 2 This assay allows high-throughput screening and aids in the diagnosis of allergies It requires only 25 L of sample and can be read on Meso Scale Diagnostics QuickPlex SQ 120 Instrument
Catalog No. 50-000-78409 Supplier Meso Scale Discovery Supplier No. K1515AR22
The R-PLEX Human IgE Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures IgE antibodies in human serum plasma or cell culture supernatant It uses electrochemiluminescence detection technology and provides quantitative results for 10 IgE biomarkers including IgE Total Ara h 1 Ara h 2 Ara h 3 Ara h 8 Ara h 9 Bos d 5 Bos d 8 Gal d 1 and Gal d 2 This assay allows high-throughput screening and aids in the diagnosis of allergies It requires only 25 L of sample and can be read on Meso Scale Diagnostics QuickPlex SQ 120 Instrument