Learn More
Meso Scale Discovery R-PLEX Human IGF-1 ASYQP
SDP
Catalog No. 5000080151
Encompass
The Meso Scale Discovery R-PLEX Human IGF-1 Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of Insulin-like Growth Factor 1 (IGF-1) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms The R-PLEX IGF-1 assay delivers precise and reproducible results for human IGF-1 quantification
Catalog No. 50-000-80151 Supplier Meso Scale Discovery Supplier No. K151AKER22
The Meso Scale Discovery R-PLEX Human IGF-1 Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of Insulin-like Growth Factor 1 (IGF-1) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L or less sample volume and can be read on MSD instrument platforms The R-PLEX IGF-1 assay delivers precise and reproducible results for human IGF-1 quantification