Learn More
Meso Scale Discovery R-PLEX Human IGFBP2 ASYQP
SDP
Catalog No. 5000078923
Encompass
The Meso Scale Discovery R-PLEX Human IGFBP-2 Assay is a multiplex immunoassay that quantitatively measures IGFBP-2 levels in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 8-logs of dynamic range and low sample volume requirements This assay allows for the accurate quantification of IGFBP-2 levels which can aid in the study of growth disorders and cancer progression With minimal hands-on time required this assay enables efficient and reproducible analysis of IGFBP-2 levels across large numbers of samples
Catalog No. 50-000-78923 Supplier Meso Scale Discovery Supplier No. K151AAVR22
Add to Cart
Add to Cart
The Meso Scale Discovery R-PLEX Human IGFBP-2 Assay is a multiplex immunoassay that quantitatively measures IGFBP-2 levels in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 8-logs of dynamic range and low sample volume requirements This assay allows for the accurate quantification of IGFBP-2 levels which can aid in the study of growth disorders and cancer progression With minimal hands-on time required this assay enables efficient and reproducible analysis of IGFBP-2 levels across large numbers of samples