Learn More
Meso Scale Discovery R-PLEX Human Ikaros ASY
SDP
Catalog No. 5000080215
Encompass
The R-PLEX Human Ikaros Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that allows for the simultaneous quantitative measurement of Ikaros protein in human serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrator The assay can be run on Meso Scale Diagnostics SECTOR Imager instrument systems and takes approximately 3 5 hours to complete
Catalog No. 50-000-80215 Supplier Meso Scale Discovery Supplier No. K1516UR2
Add to Cart
Add to Cart
The R-PLEX Human Ikaros Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that allows for the simultaneous quantitative measurement of Ikaros protein in human serum plasma and cell culture supernatants The assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrator The assay can be run on Meso Scale Diagnostics SECTOR Imager instrument systems and takes approximately 3 5 hours to complete