Learn More
Meso Scale Discovery R-PLEX Human IL-11 ASYQP
SDP
Catalog No. 5000078499
Encompass
The Meso Scale Discovery R-PLEX Human IL-11 Assay is a multiplex immunoassay panel that allows simultaneous measurement of the human cytokine Interleukin 11 (IL-11) This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements It requires only 50 L of sample per well and can be fully automated on the MSD instrument platforms The panel targets IL-11 specifically with high lot-to-lot consistency of standard curves and S/B ratios and has been validated for serum plasma and cell culture supernatants This assay allows sensitive reproducible quantitation of IL-11 levels in human samples
Catalog No. 50-000-78499 Supplier Meso Scale Discovery Supplier No. K151E2R22
The Meso Scale Discovery R-PLEX Human IL-11 Assay is a multiplex immunoassay panel that allows simultaneous measurement of the human cytokine Interleukin 11 (IL-11) This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements It requires only 50 L of sample per well and can be fully automated on the MSD instrument platforms The panel targets IL-11 specifically with high lot-to-lot consistency of standard curves and S/B ratios and has been validated for serum plasma and cell culture supernatants This assay allows sensitive reproducible quantitation of IL-11 levels in human samples