Learn More
Meso Scale Discovery R-PLEX Human IL-17RB ASY
SDP
Catalog No. 5000078921
Encompass
The R-PLEX Human IL-17RB Assay Kit from Meso Scale Discovery is a multiplex immunoassay that enables the simultaneous measurement of IL-17RB in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for the quantitative analysis of IL-17RB This assay allows for efficient and reproducible quantification of IL-17RB levels
Catalog No. 50-000-78921 Supplier Meso Scale Discovery Supplier No. K151ACUR2
The R-PLEX Human IL-17RB Assay Kit from Meso Scale Discovery is a multiplex immunoassay that enables the simultaneous measurement of IL-17RB in human serum plasma and cell culture supernatants This assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls for the quantitative analysis of IL-17RB This assay allows for efficient and reproducible quantification of IL-17RB levels