Learn More
Meso Scale Discovery R-PLEX Human IL-19 ASY
SDP
Catalog No. 5000082476
Encompass
The R-PLEX Human IL-19 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring IL-19 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies for IL-19 detection antibodies calibrators and diluents The assay takes about 3 5 hours and detects IL-19 as low as 17 pg/mL making it a sensitive and efficient tool for quantifying IL-19 in biological samples
Catalog No. 50-000-82476 Supplier Meso Scale Discovery Supplier No. K151P4R2
The R-PLEX Human IL-19 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring IL-19 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies for IL-19 detection antibodies calibrators and diluents The assay takes about 3 5 hours and detects IL-19 as low as 17 pg/mL making it a sensitive and efficient tool for quantifying IL-19 in biological samples