Learn More
Meso Scale Discovery R-PLEX Human IL-19 ASYQP
SDP
Catalog No. 5000079606
Encompass
The Meso Scale Discovery R-PLEX Human IL-19 Assay is a multiplex immunoassay panel that allows simultaneous measurement of IL-19 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent It can accurately quantify IL-19 levels with minimal hands-on time using an MSD instrument
Catalog No. 50-000-79606 Supplier Meso Scale Discovery Supplier No. K151P4R22
The Meso Scale Discovery R-PLEX Human IL-19 Assay is a multiplex immunoassay panel that allows simultaneous measurement of IL-19 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent It can accurately quantify IL-19 levels with minimal hands-on time using an MSD instrument