Learn More
Meso Scale Discovery R-PLEX Human IL-22R 2 ASY
SDP
Catalog No. 5000080386
Encompass
The Meso Scale Discovery R-PLEX Human IL-22Ralpha2 Assay is a multiplex immunoassay that measures IL-22Ralpha2 in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay can be multiplexed with other R-PLEX assays to analyze multiple biomarkers simultaneously This allows efficient and accurate quantification of IL-22Ralpha2 levels alongside other immune response markers With a testing time of just 3 5 hours this assay enables rapid profiling of IL-22Ralpha2
Catalog No. 50-000-80386 Supplier Meso Scale Discovery Supplier No. K151AAWR2
The Meso Scale Discovery R-PLEX Human IL-22Ralpha2 Assay is a multiplex immunoassay that measures IL-22Ralpha2 in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay can be multiplexed with other R-PLEX assays to analyze multiple biomarkers simultaneously This allows efficient and accurate quantification of IL-22Ralpha2 levels alongside other immune response markers With a testing time of just 3 5 hours this assay enables rapid profiling of IL-22Ralpha2