Learn More
Meso Scale Discovery R-PLEX Human IL-22R 2 ASYQP
SDP
Catalog No. 5000078957
Encompass
The Meso Scale Discovery R-PLEX Human IL-22Ralpha2 Assay is a multiplex immunoassay that measures IL-22Ralpha2 in human serum plasma and cell culture supernatant This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can be multiplexed with other R-PLEX assays to analyze multiple biomarkers simultaneously The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer The assay takes approximately 3 hours to run and only requires a small 4 L sample volume
Catalog No. 50-000-78957 Supplier Meso Scale Discovery Supplier No. K151AAWR22
The Meso Scale Discovery R-PLEX Human IL-22Ralpha2 Assay is a multiplex immunoassay that measures IL-22Ralpha2 in human serum plasma and cell culture supernatant This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay can be multiplexed with other R-PLEX assays to analyze multiple biomarkers simultaneously The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and wash buffer The assay takes approximately 3 hours to run and only requires a small 4 L sample volume