Learn More
Meso Scale Discovery R-PLEX Human IL-23R ASY
SDP
Catalog No. 5000082436
Encompass
The Meso Scale Discovery R-PLEX Human IL-23R Assay is a multiplex immunoassay for measuring IL-23R in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection on patterned arrays for high sensitivity and a wide dynamic range The kit includes a plate with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control samples for standard curve generation and assay validation It quantifies IL-23R with high precision and reproducibility suitable for 96 tests on MSD platforms
Catalog No. 50-000-82436 Supplier Meso Scale Discovery Supplier No. K151AMBR2
The Meso Scale Discovery R-PLEX Human IL-23R Assay is a multiplex immunoassay for measuring IL-23R in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection on patterned arrays for high sensitivity and a wide dynamic range The kit includes a plate with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control samples for standard curve generation and assay validation It quantifies IL-23R with high precision and reproducibility suitable for 96 tests on MSD platforms