Learn More
Meso Scale Discovery R-PLEX Human IL-23R ASYQP
SDP
Catalog No. 5000081949
Encompass
The R-PLEX Human IL-23R Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures IL-23R levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay takes 3 5 hours to complete and is suitable for measuring IL-23R during immunology and inflammation studies
Catalog No. 50-000-81949 Supplier Meso Scale Discovery Supplier No. K151AMBR22
The R-PLEX Human IL-23R Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures IL-23R levels in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay takes 3 5 hours to complete and is suitable for measuring IL-23R during immunology and inflammation studies