Learn More
Meso Scale Discovery R-PLEX Human IL-24 ASY
SDP
Catalog No. 5000079621
Encompass
The R-PLEX Human IL-24 Assay QuickPlex 5-PL Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of human Interleukin 24 (IL-24) in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides reagents for 5-plex analysis of IL-24 in a quick and streamlined workflow The assay has high sensitivity wide dynamic range and requires low sample volume This product can aid in cytokine profiling and research related to cancer and inflammatory conditions
Catalog No. 50-000-79621 Supplier Meso Scale Discovery Supplier No. K151APJR2
The R-PLEX Human IL-24 Assay QuickPlex 5-PL Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for the quantitative measurement of human Interleukin 24 (IL-24) in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides reagents for 5-plex analysis of IL-24 in a quick and streamlined workflow The assay has high sensitivity wide dynamic range and requires low sample volume This product can aid in cytokine profiling and research related to cancer and inflammatory conditions