Learn More
Meso Scale Discovery R-PLEX Human IL-24 ASYQP
SDP
Catalog No. 5000078738
Encompass
The R-PLEX Human IL-24 Assay QuickPlex 5-PL Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring human interleukin-24 (IL-24) in various biological fluids It includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator diluent The assay sensitivity is 0 3 pg/mL with a range of 0 3-2000 pg/mL requiring 25 L of sample Results are available in 3 5 hours using the QuickPlex SQ 120 instrument enabling rapid and accurate IL-24 quantification
Catalog No. 50-000-78738 Supplier Meso Scale Discovery Supplier No. K151APJR22
The R-PLEX Human IL-24 Assay QuickPlex 5-PL Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for measuring human interleukin-24 (IL-24) in various biological fluids It includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrator diluent The assay sensitivity is 0 3 pg/mL with a range of 0 3-2000 pg/mL requiring 25 L of sample Results are available in 3 5 hours using the QuickPlex SQ 120 instrument enabling rapid and accurate IL-24 quantification