Learn More
Meso Scale Discovery R-PLEX Human IL-32 ASY
SDP
Catalog No. 5000081911
Encompass
The R-PLEX Human IL-32 Assay Kit from Meso Scale Discovery measures IL-32 in human serum plasma and cell culture supernatants using electrochemiluminescence detection It offers high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with IL-32 capture antibodies detection antibodies buffers diluent and calibrators The assay takes about 3 5 hours and requires a MESO QuickPlex SQ 120 instrument It enables accurate quantitation of IL-32 an inflammatory cytokine in immune responses
Catalog No. 50-000-81911 Supplier Meso Scale Discovery Supplier No. K151AHWR2
The R-PLEX Human IL-32 Assay Kit from Meso Scale Discovery measures IL-32 in human serum plasma and cell culture supernatants using electrochemiluminescence detection It offers high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with IL-32 capture antibodies detection antibodies buffers diluent and calibrators The assay takes about 3 5 hours and requires a MESO QuickPlex SQ 120 instrument It enables accurate quantitation of IL-32 an inflammatory cytokine in immune responses