Learn More
Meso Scale Discovery R-PLEX Human IL-32 ASYQP
SDP
Catalog No. 5000081630
Encompass
The R-PLEX Human IL-32 Assay Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) for quantifying human interleukin-32 (IL-32) in various samples It employs MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection with a broad range The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD platforms and analyzed with MSD software enabling high-throughput IL-32 quantification important in inflammatory and autoimmune diseases
Catalog No. 50-000-81630 Supplier Meso Scale Discovery Supplier No. K151AHWR22
The R-PLEX Human IL-32 Assay Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) for quantifying human interleukin-32 (IL-32) in various samples It employs MSD s SULFO-TAG labels and multi-array plate technology for sensitive detection with a broad range The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD platforms and analyzed with MSD software enabling high-throughput IL-32 quantification important in inflammatory and autoimmune diseases