Learn More
Meso Scale Discovery R-PLEX Human IL-34 ASYQP
SDP
Catalog No. 5000082325
Encompass
The Meso Scale Discovery R-PLEX Human IL-34 Assay Kit is a multiplex immunoassay that measures IL-34 in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay takes 3 5 hours to complete and is suitable for measuring natural and recombinant human IL-34
Catalog No. 50-000-82325 Supplier Meso Scale Discovery Supplier No. K151R4R22
The Meso Scale Discovery R-PLEX Human IL-34 Assay Kit is a multiplex immunoassay that measures IL-34 in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay takes 3 5 hours to complete and is suitable for measuring natural and recombinant human IL-34