Learn More
Meso Scale Discovery R-PLEX Human IL-36 ASY
SDP
Catalog No. 5000082302
Encompass
The Meso Scale Discovery R-PLEX Human IL-36gamma Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of IL-36gamma in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL compound and calibrator and control solutions for generating standard curves and evaluating run validity It allows sensitive reproducible quantification of IL-36gamma levels from human samples
Catalog No. 50-000-82302 Supplier Meso Scale Discovery Supplier No. K151S8R2
The Meso Scale Discovery R-PLEX Human IL-36gamma Assay is a multiplex immunoassay panel that allows simultaneous quantitative measurement of IL-36gamma in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL compound and calibrator and control solutions for generating standard curves and evaluating run validity It allows sensitive reproducible quantification of IL-36gamma levels from human samples