Learn More
Meso Scale Discovery R-PLEX Human IL-37 ASY
SDP
Catalog No. 5000079610
Encompass
The Meso Scale Discovery R-PLEX Human IL-37 Assay is a multiplex immunoassay that measures IL-37 levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay allows for the simultaneous measurement of IL-37 and other biomarkers It requires 50 L or less of sample and can be read on the MESO QuickPlex SQ 120 instrument The assay comes with a plate coated with capture antibodies detection antibodies calibrators and controls
Catalog No. 50-000-79610 Supplier Meso Scale Discovery Supplier No. K151AJNR2
The Meso Scale Discovery R-PLEX Human IL-37 Assay is a multiplex immunoassay that measures IL-37 levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This assay allows for the simultaneous measurement of IL-37 and other biomarkers It requires 50 L or less of sample and can be read on the MESO QuickPlex SQ 120 instrument The assay comes with a plate coated with capture antibodies detection antibodies calibrators and controls