Learn More
Meso Scale Discovery R-PLEX Human IL-37 ASYQP
SDP
Catalog No. 5000079130
Encompass
The R-PLEX Human IL-37 Assay from Meso Scale Discovery is a multiplex immunoassay that measures IL-37 in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of IL-37 an anti-inflammatory cytokine with a run time of approximately 2 5 hours It includes a plate coated with capture antibodies detection antibodies labeled with an ECL compound and calibrator and control solutions
Catalog No. 50-000-79130 Supplier Meso Scale Discovery Supplier No. K151AJNR22
The R-PLEX Human IL-37 Assay from Meso Scale Discovery is a multiplex immunoassay that measures IL-37 in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the quantitative measurement of IL-37 an anti-inflammatory cytokine with a run time of approximately 2 5 hours It includes a plate coated with capture antibodies detection antibodies labeled with an ECL compound and calibrator and control solutions