Learn More
Meso Scale Discovery R-PLEX Human IL1RL1/ST2 ASY
SDP
Catalog No. 5000080282
Encompass
The R-PLEX Human IL1RL1/ST2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures IL1RL1/ST2 in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound and calibrators and controls to generate a standard curve The assay takes 3 5 hours to complete and requires a Meso Scale Discovery instrument such as SECTOR Imager QuickPlex SQ 120 or QuickPlex SQ 120
Catalog No. 50-000-80282 Supplier Meso Scale Discovery Supplier No. K1514HR2
The R-PLEX Human IL1RL1/ST2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures IL1RL1/ST2 in human serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound and calibrators and controls to generate a standard curve The assay takes 3 5 hours to complete and requires a Meso Scale Discovery instrument such as SECTOR Imager QuickPlex SQ 120 or QuickPlex SQ 120