Learn More
Meso Scale Discovery R-PLEX Human IRS-1 ASY
SDP
Catalog No. 5000082446
Encompass
The R-PLEX Human IRS-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human IRS-1 in cell and tissue lysates It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay has high specificity and low cross-reactivity It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator The kit can test up to 80 samples in duplicate and has a testing time of 2 5 hours It is for research use only
Catalog No. 50-000-82446 Supplier Meso Scale Discovery Supplier No. K1517DR2
The R-PLEX Human IRS-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human IRS-1 in cell and tissue lysates It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay has high specificity and low cross-reactivity It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator The kit can test up to 80 samples in duplicate and has a testing time of 2 5 hours It is for research use only