Learn More
Meso Scale Discovery R-PLEX Human Jagged-1 ASY
SDP
Catalog No. 5000082061
Encompass
The R-Plex Human Jagged-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that enables the simultaneous measurement of Jagged-1 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized calibrator and controls to generate a standard curve and validate assay performance The assay has a typical lower limit of detection of 1 5 pg/mL and an upper limit of quantitation of 1500 pg/mL for Jagged-1
Catalog No. 50-000-82061 Supplier Meso Scale Discovery Supplier No. K151AMCR2
The R-Plex Human Jagged-1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that enables the simultaneous measurement of Jagged-1 in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires a low sample volume The kit includes a 96-well plate pre-coated with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized calibrator and controls to generate a standard curve and validate assay performance The assay has a typical lower limit of detection of 1 5 pg/mL and an upper limit of quantitation of 1500 pg/mL for Jagged-1