Learn More
Meso Scale Discovery R-PLEX Human Klotho ASYQP
SDP
Catalog No. 5000079428
Encompass
The Meso Scale Discovery R-PLEX Human Klotho Assay is a multiplex immunoassay for measuring Klotho protein in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay quantifies both secreted and membrane-bound Klotho using two monoclonal antibodies providing results in 3 5 hours With a detection limit of 19 5 fg/mL it measures Klotho levels across various concentrations crucial for studying aging and age-related diseases
Catalog No. 50-000-79428 Supplier Meso Scale Discovery Supplier No. K151AAXR22
The Meso Scale Discovery R-PLEX Human Klotho Assay is a multiplex immunoassay for measuring Klotho protein in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay quantifies both secreted and membrane-bound Klotho using two monoclonal antibodies providing results in 3 5 hours With a detection limit of 19 5 fg/mL it measures Klotho levels across various concentrations crucial for studying aging and age-related diseases