Learn More
Meso Scale Discovery R-PLEX Human LBP ASYQP
SDP
Catalog No. 5000079953
Encompass
The R-PLEX Human LBP Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the quantitative measurement of lipopolysaccharide binding protein (LBP) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls The assay can detect LBP in the range of 0 05-400 ng/mL and requires only 25 L of sample
Catalog No. 50-000-79953 Supplier Meso Scale Discovery Supplier No. K151K5R22
Add to Cart
Add to Cart
The R-PLEX Human LBP Assay from Meso Scale Diagnostics is a multiplex immunoassay kit that allows for the quantitative measurement of lipopolysaccharide binding protein (LBP) in human serum plasma and cell culture supernatants This assay utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer sample diluent and calibrators and controls The assay can detect LBP in the range of 0 05-400 ng/mL and requires only 25 L of sample