Learn More
Meso Scale Discovery R-PLEX Human LIF ASY
SDP
Catalog No. 5000081928
Encompass
The R-PLEX Human LIF Assay Sector 5-PL is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the simultaneous quantitative measurement of Lif in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run on Meso Scale Diagnostics SECTOR Imager 2400 or 6000 instruments and results are obtained in as little as 3 5 hours Each kit provides reagents for 96 tests in a convenient plate format
Catalog No. 50-000-81928 Supplier Meso Scale Discovery Supplier No. K151AJPR2
The R-PLEX Human LIF Assay Sector 5-PL is a multiplex immunoassay kit from Meso Scale Diagnostics that allows for the simultaneous quantitative measurement of Lif in human serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run on Meso Scale Diagnostics SECTOR Imager 2400 or 6000 instruments and results are obtained in as little as 3 5 hours Each kit provides reagents for 96 tests in a convenient plate format