Learn More
Meso Scale Discovery R-PLEX Human LRRK2 ASY
SDP
Catalog No. 5000080606
Encompass
The R-Plex Human LRRK2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring total and phosphorylated LRRK2 in human samples It uses electrochemiluminescence detection for sensitive quantitative monitoring of LRRK2 expression and phosphorylation at Ser935 Ser910 and Ser973 The kit includes a 96-well plate with capture and detection antibodies calibrator diluent wash buffer read buffer and a protocol Sensitivity is 0 3-0 8 pg/mL requiring 50 L of sample per well suitable for human cell lysates cerebrospinal fluid serum plasma and other samples
Catalog No. 50-000-80606 Supplier Meso Scale Discovery Supplier No. K1511PR2
The R-Plex Human LRRK2 Assay Kit from Meso Scale Discovery is a multiplex immunoassay for measuring total and phosphorylated LRRK2 in human samples It uses electrochemiluminescence detection for sensitive quantitative monitoring of LRRK2 expression and phosphorylation at Ser935 Ser910 and Ser973 The kit includes a 96-well plate with capture and detection antibodies calibrator diluent wash buffer read buffer and a protocol Sensitivity is 0 3-0 8 pg/mL requiring 50 L of sample per well suitable for human cell lysates cerebrospinal fluid serum plasma and other samples