Learn More
Meso Scale Discovery R-PLEX Human LRRK2 ASYQP
SDP
Catalog No. 5000078389
Encompass
The R-PLEX Human LRRK2 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures the levels of total LRRK2 and phosphorylated LRRK2 at serines 935 1297 and 1292 in human biological samples such as serum plasma and cell lysates The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrator diluent wash buffer read buffer and a quick guide protocol This assay allows for the efficient analysis of LRRK2 protein levels and phosphorylation
Catalog No. 50-000-78389 Supplier Meso Scale Discovery Supplier No. K1511PR22
The R-PLEX Human LRRK2 Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that measures the levels of total LRRK2 and phosphorylated LRRK2 at serines 935 1297 and 1292 in human biological samples such as serum plasma and cell lysates The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrator diluent wash buffer read buffer and a quick guide protocol This assay allows for the efficient analysis of LRRK2 protein levels and phosphorylation