Learn More
Meso Scale Discovery R-PLEX Human LRRK2 pS935 ASY
SDP
Catalog No. 5000080554
Encompass
The R-Plex Human LRRK2 pS935 Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of LRRK2 phosphorylated at serine 935 in human biological samples such as cell lysates tissue homogenates and other sample types The assay utilizes an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and positive control The assay can be run on Meso Scale Discovery instrumentation including the QuickPlex SQ 120
Catalog No. 50-000-80554 Supplier Meso Scale Discovery Supplier No. K1511QR2
The R-Plex Human LRRK2 pS935 Assay Kit from Meso Scale Discovery is used to quantitatively measure levels of LRRK2 phosphorylated at serine 935 in human biological samples such as cell lysates tissue homogenates and other sample types The assay utilizes an electrochemiluminescent detection method and provides high sensitivity broad dynamic range and rapid results The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator and positive control The assay can be run on Meso Scale Discovery instrumentation including the QuickPlex SQ 120