Learn More
Meso Scale Discovery R-PLEX Human Lymphotactin ASY
SDP
Catalog No. 5000080362
Encompass
The R-PLEX Human Lymphotactin Assay by Meso Scale Discovery is a multiplex immunoassay for measuring lymphotactin in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity up to 8-logs of dynamic range and low sample volume needs It quantifies lymphotactin as low as 0 04 pg/mL with precision The assay allows simultaneous measurement of lymphotactin and other biomarkers providing rapid results in hours The kit includes a 96-well plate detection antibodies calibrators controls buffers plate seals and an MSD instrument
This item is not returnable. View return policy
Catalog No. 50-000-80362 Supplier Meso Scale Discovery Supplier No. K151ACVR2
The R-PLEX Human Lymphotactin Assay by Meso Scale Discovery is a multiplex immunoassay for measuring lymphotactin in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity up to 8-logs of dynamic range and low sample volume needs It quantifies lymphotactin as low as 0 04 pg/mL with precision The assay allows simultaneous measurement of lymphotactin and other biomarkers providing rapid results in hours The kit includes a 96-well plate detection antibodies calibrators controls buffers plate seals and an MSD instrument