Learn More
Meso Scale Discovery R-PLEX Human MAdCAM-1 ASY
SDP
Catalog No. 5000078959
Encompass
The R-PLEX Human MAdCAM-1 Assay is an ELISA-based kit for quantifying MAdCAM-1 in human serum plasma and other fluids It uses a sandwich enzyme immunoassay with a monoclonal antibody pre-coated on a microplate Samples bind to the antibody and after washing a biotinylated detection antibody is added Following another wash HRP conjugate is added A substrate solution develops color proportional to MAdCAM-1 levels which is then measured The assay offers high sensitivity and specificity for MAdCAM-1 detection
Catalog No. 50-000-78959 Supplier Meso Scale Discovery Supplier No. K151AAYR2
The R-PLEX Human MAdCAM-1 Assay is an ELISA-based kit for quantifying MAdCAM-1 in human serum plasma and other fluids It uses a sandwich enzyme immunoassay with a monoclonal antibody pre-coated on a microplate Samples bind to the antibody and after washing a biotinylated detection antibody is added Following another wash HRP conjugate is added A substrate solution develops color proportional to MAdCAM-1 levels which is then measured The assay offers high sensitivity and specificity for MAdCAM-1 detection