Learn More
Meso Scale Discovery R-PLEX Human Mesothelin ASY
SDP
Catalog No. 5000081687
Encompass
The R-PLEX Human Mesothelin Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human mesothelin in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity a wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD ECL-capable instrument enabling accurate mesothelin quantification a key biomarker for certain cancers and diseases
Catalog No. 50-000-81687 Supplier Meso Scale Discovery Supplier No. K1510HR2
The R-PLEX Human Mesothelin Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human mesothelin in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity a wide dynamic range and low sample volume The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and necessary reagents Results are available in 3 5 hours using an MSD ECL-capable instrument enabling accurate mesothelin quantification a key biomarker for certain cancers and diseases