Learn More
Meso Scale Discovery R-PLEX Human MET soluble ASY
SDP
Catalog No. 5000078893
Encompass
The R-PLEX Human Met Soluble Assay from Meso Scale Discovery is a multiplex immunoassay that measures 10 metabolic biomarkers in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The panel includes insulin leptin PAI-1 total resistin MCP-1 adiponectin TNF-alpha IL-6 CRP and visfatin The assay provides efficient analysis of metabolic and inflammatory status with a protocol time of just over 3 hours It includes a 96-well plate with capture antibodies detection antibodies and calibrators For research use only
Catalog No. 50-000-78893 Supplier Meso Scale Discovery Supplier No. K1510ZR2
The R-PLEX Human Met Soluble Assay from Meso Scale Discovery is a multiplex immunoassay that measures 10 metabolic biomarkers in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The panel includes insulin leptin PAI-1 total resistin MCP-1 adiponectin TNF-alpha IL-6 CRP and visfatin The assay provides efficient analysis of metabolic and inflammatory status with a protocol time of just over 3 hours It includes a 96-well plate with capture antibodies detection antibodies and calibrators For research use only